Learn More
Enterprise Technology Solutions Genuine Joe Bottle, 16oz, 24/CT, Translucent
SDP
Supplier:  Enterprise Technology Solutions GJO85153
Encompass_Preferred
Plastic bottle with a 28mm/400 neck finish is made of polyethylene to safely contain a wide variety of liquids. Convenient graduation marks allow you to make quick measurements inside the bottle. Clear, translucent bottle lets you easily monitor the current level of liquid contained.
Catalog No. 50-008-11235