Learn More
Enterprise Technology Solutions Genuine Joe Floor Pads, f/Heavy-duty Stripping, 16", 5/CT, Black
SDP
Supplier:  Enterprise Technology Solutions GJO90216
Encompass_Preferred
Heavy-duty floor stripping pad works with your stripping solution and floor machine agitation to remove all finish from floors. Open-web design allows dirt to enter the fiber and resist soil load on the face of the pad for maximum use. Resiliency allows pad to remain flexible to reach depressions. Heavier denier fibers mean longer-lasting pads. Harder resin is abrasive for aggressive stripping action. Floor pad is designed for use at speeds between 175 to 350 RPM in rotary/automatic scrubbers.
Catalog No. 50-008-11061