Learn More
Enterprise Technology Solutions Genuine Joe Utility Brush, w/Eyelet, Nylon, 9" Handle, White
SDP
Supplier:  Enterprise Technology Solutions GJO98215
Encompass_Preferred
This Nylon Utility Brush features silky, flexible synthetic fiber with superior abrasion-resistance. The fibers are also heat-resistant. Brush stands up to common cleaners, detergents, acids or alkalis, and has excellent water-retention. It does not mat or hold odors. Eyelet in the easy-to-grip, 9" handle allows easy hanging. Structural foam block offers a reliable construction. Brush is recommended for general cleaning in foodservice and institutional environments.
Catalog No. 50-008-31063