Learn More
Medchemexpress LLC Geri-BP002-A, novel inhibitor of acyl-CoA: cholesterol acyltransferase produced by Aspergillus fumigatus F93 | 119-47-1 | 99.8% | 25 G
SDP
Supplier:  Medchemexpress LLC HY12125425G
Encompass_Preferred
An inhibitor of acyl-CoA:cholesterol acyltransferase (ACAT) produced by Aspergillus fumigatus F93, supplied as a solid research reagent for cancer and cytotoxicity studies.
  • Exhibits cytotoxicity against A549, SKOV-3, HCT-15, XF-498, and SKMEL-2 cells.
  • Has ED50 values of 5.45 μg/mL for A549, 8.83 μg/mL for SKOV-3, 5.79 μg/mL for HCT-15, 7.30 μg/mL for XF-498, and 5.39 μg/mL for SKMEL-2.
  • Available as solid in multiple package sizes and as a 10 mM solution in DMSO.
Catalog No. 50-002-32527