Learn More
Cayman Chemical GermIcIdIn A 5mg
SDP
Supplier:  Cayman Chemical 199715 MG
Encompass_Preferred
An autoregulative inhibitor of spore germination in the genus Streptomyces at concentrations as low as 200 pM; also inhibits the porcine Na+/K+-activated ATPase (ID50 = 100 µM)
Catalog No. 50-298-2628