Learn More
Cayman Chemical Gernylgernylaceton 10mg
SDP
Supplier:  Cayman Chemical 2021810 MG
Encompass_Preferred
An inducer of Hsp expression that increases Hsp70, Hsp22, Hsp27, Hsp90, and Hsp105 levels in various cells and tissues; orally bioavailable and has diverse effects in vivo, including hepatoprotective, neuroprotective, and antiulcerative effects
Catalog No. 50-298-4578