Learn More
Apexbio Technology LLC Gestodene 60282-87-3 10mg
SDP
Supplier:  Apexbio Technology LLC B151710
Encompass_Preferred
Gestodene (60282-87-3) is a small-molecule agonist targeting progesterone receptors It is designed to modulate the hypothalamic-pituitary-gonadal axis thereby regulating hormonal secretion and ovulatory processes Gestodene exerts its biological activity primarily through high-affinity binding and agonist activity on progesterone receptors leading to suppression of luteinizing hormone (LH) and follicle-stimulating hormone (FSH) secretion inhibition of ovulation alteration of cervical mucus viscosity and modification of endometrial morphology Based on these pharmacological properties Gestodene holds research potential in reproductive biology endocrinology and contraceptive mechanism studies
Catalog No. 50-000-92210