Learn More
Medchemexpress LLC HY-101405 10mg Medchemexpress, Gestrinone CAS:16320-04-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10140510MG
Encompass_Preferred
Medchemexpress, HY-101405 10mg Gestrinone CAS:16320-04-0 Gestrinone (R2323) is a synthetic steroid hormone used to treat endometriosis. It inhibits leiomyoma cells with an IC 50 of 43.67 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2112