Learn More
MSC GF Gage Go/No Go Threaded Ring Gage, #12-24 UNC, Class 2A
Supplier:  MSC 55325252
Encompass
This threaded ring gage is designed for precisely inspecting external threads. It features both a Go and No Go gaging system for comprehensive thread verification, ensuring components meet exact specifications.
  • For #12-24 UNC threads.
  • Manufactured to Class 2A tolerance.
  • Provides Go and No Go gaging.
  • Double ring configuration for ease of use.
  • Made in the United States.
Catalog No. 50-005-27807