Learn More
MSC GF Gage Plug Thread Gage, 1/2-13 Thread, 2B & 3B Class, Single End, Go, Titanium Nitride
Supplier:  MSC 55313019
Encompass
This single-end plug thread gage is designed for accurately checking 1/2-13 threads. It is designated as a 'Go' gage, suitable for both 2B and 3B thread classes. Manufactured from High-Speed Steel and featuring a Titanium Nitride coating, this gage provides reliable performance with a Go Pitch Diameter of 0.4500 inches.
  • Designed for 1/2-13 threads
  • Supports 2B and 3B thread classes
  • Single-end 'Go' gage type
  • Features a Titanium Nitride coating
  • Constructed from high-speed steel
  • Go pitch diameter of 0.4500 inches
  • Does not include a handle
Catalog No. 50-005-30412