Learn More
MSC GF Gage Plug Thread Gage, #10-32 Thread, 3B Class, Double End, Go/No Go
Supplier:  MSC 55311054
Encompass
This plug thread gage is designed for precise measurement of internal threads. It features a #10-32 thread, 3B class, and a double-end design with both Go and No Go ends for accurate tolerance checking. Constructed from hardened tool steel, it ensures durability and reliability in demanding applications.
  • #10-32 thread for specific applications.
  • 3B class for precise thread measurement.
  • Double-end design with both Go and No Go ends for comprehensive checking.
  • Constructed from hardened tool steel for durability.
  • Includes a handle for ease of use.
  • Manufactured in the United States.
Catalog No. 50-005-27091