Learn More
MSC GF Gage Plug Thread Gage, 3/4-16 Thread, 2B Class, No Go
Supplier:  MSC 55308076
Encompass
This plug thread gage features a 3/4-16 thread and 2B class, designed for precision measurement with a single end and a No Go pitch diameter of 0.7159 inches.
  • No Go design for accurate thread inspection
  • 3/4-16 thread size
  • Constructed from hardened tool steel
  • 2B class for specified thread tolerance
  • No Go pitch diameter of 0.7159 inch
  • Single end configuration
  • Made in the United States
Catalog No. 50-005-28208