Learn More
MSC GF Gage Plug Thread Gage, #6-32 Thread, 2B Class, Double End, Go & No Go
Supplier:  MSC 05356332
Encompass
This plug thread gage is specifically designed for accurately measuring #6-32 threads. It features a double-ended design, incorporating both "Go" and "No Go" sides to facilitate comprehensive thread inspection. Manufactured from hardened tool steel, this gage ensures durability and precise 2B class fit measurements, making it suitable for various industrial applications.
  • Designed for measuring #6-32 threads
  • Double-ended design for comprehensive inspection
  • Features both "Go" and "No Go" sides
  • Ensures 2B class fit accuracy
  • Constructed from hardened tool steel for durability
  • Includes a handle for ease of use
  • Go pitch diameter: 0.1177 inch
  • No Go pitch diameter: 0.12140 inch
Catalog No. 50-005-29628