Learn More
MSC GF Gage Plug Thread Gage Handle, 5 Handle Size, Steel
Supplier:  MSC 55338610
Encompass
This plug thread gage handle is designed to accommodate various plug thread sizes, including 1 - 11-1/2, 1-1/2 - 11-1/2, 1-1/4 - 11-1/2, and 2 - 11-1/2. Constructed from steel, it provides a durable solution for gaging applications.
  • Constructed from steel for durability
  • Accommodates plug thread sizes 1 - 11-1/2, 1-1/2 - 11-1/2, 1-1/4 - 11-1/2, and 2 - 11-1/2
  • Features a size 5 handle
Catalog No. 50-005-28843