Learn More
MSC GF Gage Plug Thread Gage, M3x0.50 Thread, 6H Class, Single End, Go
Supplier:  MSC 55318430
Encompass
This plug thread gage is designed for precise measurement of internal threads. It features an M3x0.50 thread size and 6H classification, functioning as a single-end 'Go' gage to verify that the thread meets the minimum material limit. Constructed from hardened tool steel, this gage ensures durability and accuracy in quality control applications.
  • Go plug thread gage for internal thread verification
  • M3x0.50 thread size for specific applications
  • Constructed from hardened tool steel for extended durability
  • 6H classification ensures adherence to international standards
  • Features a 2.6800 mm go pitch diameter
  • Single end design for focused measurement
  • Handle is not included, allowing for custom handle integration
Catalog No. 50-005-30372