Learn More
MSC GF Gage Plug Thread Gage, M5x0.80 Thread, 6H Class, No Go, Single End
Supplier:  MSC 55318851
Encompass
This Plug Thread Gage features an M5x0.80 thread and a 6H class. It is a single-end, No Go type gage, with a No Go Pitch Diameter of 0.1813 inches. The gage is made from hardened tool steel and is manufactured in the United States.
  • No go thread gage
  • M5x0.8 thread size
  • 6H class
  • Single end design
  • Made from hardened tool steel
  • No go pitch diameter: 0.18130 in (4.6100 mm)
  • Handle not included
  • Manufactured in the United States
Catalog No. 50-005-28137