Learn More
MSC GF Gage Threaded Ring Gage, 1/2-13 Thread, UNC, Class 3A, Go
Supplier: MSC 55322051
This threaded ring gage is designed for precise measurement of 1/2-13 UNC Class 3A 'Go' threads. It is manufactured by a leading supplier of precision thread gages, utilizing advanced MasterScanners for accuracy and can feature a TiN coating for enhanced durability and performance in demanding applications.
- Designed for precise measurement of 1/2-13 UNC Class 3A 'Go' threads.
- Manufactured using advanced MasterScanners for high accuracy.
- Offers optional TiN coating for additional hardness and lubricity.
- Enhances gage life and improves surface hardness.
- Provides corrosion and abrasion resistance.
- Suitable for severe gaging applications or high-volume product inspection.