Learn More
MSC GF Gage Threaded Ring Gage, 1/2-20 UNF, 2A, Go/No Go
Supplier:  MSC 55325559
Encompass
The Threaded Ring Gage is designed for precise measurement of threads. This particular model features a 1/2-20 UNF thread size with a 2A class, functioning as both a "Go" and "No Go" gage. It is a double-ring type, ensuring comprehensive thread inspection. Manufactured in the United States, it comes standard with conformance certification, and NIST traceable long-form certification can be requested.
  • Comes standard with conformance certification; NIST traceable long-form certification available upon request.
  • Double-ring design for comprehensive thread inspection.
  • Hassle-free guarantee allows for exchange or refund within 30 days of shipment.
Catalog No. 50-005-27645