Learn More
MSC GF Gage Threaded Ring Gage, #5-44 Thread, UNF, Class 2A, Go
Supplier:  MSC 55320477
Encompass
This threaded ring gage is designed for accurately checking #5-44 UNF 2A threads. It functions as a 'Go' gage, ensuring the thread is not undersized, and is made from durable tool steel for reliable performance.
  • Measures #5-44 thread size.
  • Features UNF thread type.
  • Ensures 2A classification.
  • Functions as a "Go" gage.
  • Designed as a single ring.
  • Constructed from tool steel.
Catalog No. 50-005-27816