Learn More
MSC GF Gage Threaded Ring Gage, 7/8-9 Thread, UNC, Class 2A, Go/No Go
Supplier: MSC 55325773
This threaded ring gage provides precise measurement and inspection of 7/8-9 UNC external threads. Designed with both Go and No Go gages, it ensures that components meet the specified 2A thread classification for proper fit and functionality in various applications.
- Features both Go and No Go gages for comprehensive thread inspection.
- Specifically designed for 7/8-9 UNC thread configurations.
- Conforms to 2A thread classification standards.
- Double ring design enhances usability and accuracy.