Learn More
MSC GF Gage Threaded Ring Gage, 9/16-24, UNEF, 2A, Go & No Go
Supplier:  MSC 55325633
Encompass
This threaded ring gage offers precise measurement capabilities for quality control and inspection. It is engineered with specific thread characteristics to ensure accurate assessment of threaded components.
  • Designed for 9/16-24 thread size.
  • Features UNEF thread type.
  • Classified as 2A for thread accuracy.
  • Includes both go and no go elements for comprehensive inspection.
  • Double ring design for ease of use.
  • Manufactured in the United States.
Catalog No. 50-005-30065