Learn More
MSC GF Gage Truncated Taperlock Gage, #10-32 UNF, Go & No Go, 2A
Supplier:  MSC 55332191
Encompass
The GF Gage Truncated Taperlock Gage is designed for precise measurement of #10-32 UNF threads. This steel gage features both 'Go' and 'No Go' sides to ensure accurate thread inspection, providing a medium fit with its 2A classification. It comes complete with a handle for ease of use and includes a Certificate of Conformance.
  • Designed for #10-32 UNF threads
  • Features go and no go functionality
  • Constructed from durable steel
  • Includes a handle for convenient use
  • Provides a medium fit with 2A classification
  • Comes with a certificate of conformance
Catalog No. 50-005-27652