Learn More
MSC GF Gage Truncated Taperlock Gage, M2.5 x 0.45, No Go, Class 6G
Supplier:  MSC 55335632
Encompass
This truncated taperlock gage is engineered for precise thread inspection. It features a specific thread size of M2.5 x 0.45 and functions as a 'No Go' gage, ensuring that threads exceeding the maximum material limit are accurately identified. Meeting the rigorous 6G classification, this gage is suitable for demanding metalworking applications. Its manufacturing process incorporates advanced techniques, including MasterScanners, to guarantee exceptional accuracy and reliability.
  • Truncated taperlock design for thread gaging
  • No go functionality to check oversized threads
  • M2.5 x 0.45 thread size for specific applications
  • 6G classification for high-precision measurement
  • Produced using advanced MasterScanners for superior accuracy
Catalog No. 50-005-30418