Learn More
MSC GF Gage Truncated Taperlock Thread Gage, M5x0.8, No Go, Class 6G
Supplier:  MSC 55335715
Encompass
This Truncated Taperlock Thread Gage is engineered for precise inspection of M5x0.8 internal threads. It functions as a 'No Go' gage, ensuring that threads do not exceed the upper limit of the specified tolerance, critical for quality control in manufacturing and assembly processes.
  • Truncated Taperlock design for accurate thread gaging
  • M5x0.8 thread size for specific application needs
  • No Go classification to verify maximum thread size limits
  • Class 6G for adherence to specific tolerance standards
  • Manufactured in the United States
Catalog No. 50-005-30679