Learn More
Foxx Life Sciences EZLabpure™ High Particulate Syringe Filter, 0.45µm Hydrophilic PVDF membrane with 1µm Glass Fiber Prefilter with Female Luer-Lok™ Inlet and Male Luer Slip, Diameter 25mm, Non-Sterile, 100/PK
Supplier:  Foxx Life Sciences 74P3216FLS
Encompass_Preferred
EZLabpure PVDF Syringe Filters with Glass Fiber are designed for demanding laboratory filtration tasks, providing reliable performance for high particulate loads.
Available in 0.22 um pore size with 25 mm diameter, these filters are widely used in environmental testing, air monitoring, and other prefiltration workflows.
The glass fiber membrane captures large debris and turbidity, extending filter life and maintaining throughput for challenging samples.
Each filter is built with polypropylene housing and fitted with a female Luer-Lok inlet and male Luer slip outlet to ensure secure, leak-free operation. Manufactured from USP Class compliant materials, these filters deliver durability and consistency in routine and high-demand laboratory applications.
Features
High capacity for particulate-rich samples reduces clogging
Extended membrane life supports efficient sample throughput
Bulk pack format enhances laboratory convenience
Catalog No. 50-000-87440