Learn More
Medchemexpress LLC GFB-024 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9905901MG
Encompass_Preferred
GFB-024 is a CHO-expressed humanized antibody that targets CB1/CNR1 GFB-024 has a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for GFB-024 can refer to Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24778