Learn More
Darwin Chambers Company GFCI PROTECTED DUPLEX OUTLET
SDP
Supplier:  Darwin Chambers Company RMDUPLEX
Encompass_Preferred
GFCI protected duplex outlet, foamed in place, located per customer instruction. Darwin Chambers to provide outlet, splash-proof in-use cover, thermal protection cut-off, and wiring from outlet to top of room. Wiring to junction boxes on top of room and circuiting by others. Please note: at present, available duplex outlets have a rating of -40C to 60C.
Catalog No. NC1587926