Learn More
MSC GF Gage Truncated Taperlock Thread Gage, #6-32, No Go, Class 2A, 1 ea
Supplier:  MSC 55329775
Encompass
The truncated taperlock thread gage is a precision instrument designed for verifying the accuracy of internal threads. This No Go gage is engineered to ensure that thread dimensions meet strict quality standards for manufacturing and inspection applications.
  • Designed as a truncated taperlock thread gage
  • Features a #6-32 thread size
  • Class 2A classification
  • Constructed from durable steel
  • Designed for No Go inspection processes
Catalog No. 50-005-11538