Learn More
MSC GF Gage Truncated Taperlock Thread Setting Plug Gage, 5/16-18 UNC, 3A, Steel, 1 ea.
Supplier:  MSC 55333470
Encompass
This truncated taperlock thread gage is designed for inspecting the accuracy of internal threads. It features a go and no go configuration for reliable thread verification, constructed from steel for durability.
  • Thread size 5/16-18 UNC
  • Classification 3A
  • Truncated taperlock style
  • Includes handle
  • Made in the United States
Catalog No. 50-005-47970