Learn More
Santa Cruz Biotechnology GFP Antibody (B-2) HRP, 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC9996 HRP
Encompass
GFP Antibody (B-2) is a high quality monoclonal GFP antibody (also designated Green Fluorescent Protein antibody) suitable for the detection of the GFP protein. GFP Antibody (B-2) is available as both the non-conjugated anti-GFP antibody form, as well as multiple conjugated forms of anti-GFP antibody, including agarose, HRP, PE, FITC and multiple Alexa Fluor conjugates.
Catalog No. NC1703812