Learn More
Proteintech Group Inc GFP-Trap Agarose, kit
SDP
Supplier:  Proteintech Group Inc GTAK20
Encompass
GFP-Trap Agarose is an affinity resin for IP of GFP-fusion proteins. It consists of a GFP Nanobody/ VHH coupled to agarose beads. The ChromoTek GFP-Trap Agarose is a ready-to-use reagent.
Catalog No. 50-225-8333