Learn More
Aviva Systems Biology Corp GFRA4 Antibody - C-terminal region (ARP61585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GFRA4. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-411