Learn More
Medchemexpress LLC Ggfg-amide-glycol-amide-exatecan | 2866301-18-8 | 98.7% | 870.88 | C43H47FN8O11 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1536001MG
Encompass_Preferred
GGFG-amide-glycol-amide-Exatecan is a synthetic intermediate derived from exatecan used in the synthesis of antibody-drug conjugates (ADCs) for research applications. It is provided for research use only.
  • Intermediate for exatecan-derived ADC payload synthesis
  • Suitable for linker and conjugation chemistry
  • Reported purity 98.7%
  • Available in milligram quantities for medicinal chemistry
Catalog No. 50-002-04353