Learn More
Medchemexpress LLC Ggfg-amide-glycol-amide-exatecan | 2866301-18-8 | 98.7% | 870.88 | C43H47FN8O11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1536005MG
Encompass_Preferred
GGFG-amide-glycol-amide-Exatecan is an Exatecan derivative used as an intermediate in the synthesis of antibody-drug conjugates (ADCs). Supplied for research use only, it is provided in small-quantity packages suitable for synthetic and analytical workflows.
  • High purity (98.7%) suitable for research applications.
  • Molecular formula C43H47FN8O11, molecular weight 870.88.
  • Used as a drug-linker intermediate in ADC synthesis.
  • Available in small pack sizes (1 mg-100 mg) to support development and scale-up.
  • Provided with a datasheet and SDS for handling and safety information.
Catalog No. 50-002-04485