Learn More
Medchemexpress LLC GGFG-amide-glycol-amide-Exatecan | 2866301-18-8 | 98.7% | 870.88 g/mol | C43H47FN8O11 | 25 MG

Supplier: Medchemexpress LLC HY15360025MG
GGFG-amide-glycol-amide-Exatecan is an Exatecan derivative used as an intermediate in the synthesis of antibody-drug conjugates (ADCs). It is supplied as a high-purity solid with documented analytical data and formulation guidance for research use.
- High purity: 98.7%.
- Solid, off-white to light yellow appearance.
- Molecular weight 870.88 g/mol.
- Soluble in DMSO (100 mg/mL); in vivo formulation protocols provided.
- Storage guidance for solvent and sealed material to preserve stability.
- Analytical documentation available: COA, HNMR, LCMS, SDS.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.