Learn More
AnaSpec GIP (1-42), human, 1mg
Supplier:  AnaSpec AS612261
Encompass
GIP (1-42), human[YAEGTFISDYSIAMDKIHQQDFVNWLLAQKGKKNDWKHNITQ] ; MW 4983.6; (1 mg) GIP (Glucose-dependent Insulinotropic Polypeptide or also known as Gastric Inhibitory Polypeptide) is a 42-amino acid peptide released by the K cells of the duodenum and jejunum in response to food intake. GIP, together with GLP (Gastric-like Peptide) are members of the hormone peptide family of Incretins which stimulate insulin secretion from pancreatic islet B -cells, and also appears to promote beta cell proliferation and beta cell survival. Recent studies suggest that GIP plays a role in lipid homeostasis and possibly in the pathogenesis of obesity.
Catalog No. NC0554104