Learn More
Gold Biotechnology Inc Gibberellic Acid 3
SDP
Supplier:  Gold Biotechnology Inc G12025
Encompass_Preferred
Quick-Dissolve Gibberellic Acid is a special formulation developed at GoldBio that dissolves more easily in water than standard gibberellic Acid In our lab KOH is not required to drive the Quick-Dissolve Gibberellic Acid into solution
Catalog No. 50-005-62091