Learn More
Selleck Chemical LLC Gibberellic acid S4766-25mg
SDP
Supplier:  Selleck Chemical LLC S476625MG
Encompass_Preferred
Gibberellic acid (Gibberellin Gibberellin A3 GA3 GA) a plant hormone stimulating plant growth and development is a tetracyclic di-terpenoid compound
Catalog No. 50-310-9371