Learn More
Medchemexpress LLC Gibberellic acid | 77-06-5 | 346.37 | 1 G
SDP
Supplier:  Medchemexpress LLC HYN19641G
Encompass_Preferred
Gibberellic acid, named after the fungus Gibberella fujikuroi, regulates crucial processes in plant development and growth, including seed development, germination, stem and root growth, cell division, and flowering time. It also enhances plant resilience to various environmental stresses such as cold, drought, and heavy metals. Research indicates its involvement in increased lipid peroxidation and fluctuations in the antioxidant defense system in rats.
  • Regulates plant development and growth
  • Improves plant response to environmental stresses
  • Solid, white to off-white appearance
  • Soluble in DMSO and H2O
  • Stable under recommended storage conditions
Catalog No. 50-004-15819