Learn More
Medchemexpress LLC Gibberellic acid | 77-06-5 | 346.37 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN1964500MG
Encompass_Preferred
Gibberellic acid, also known as Gibberellin A3, is a phytohormone that plays a crucial role in plant development and growth processes. It regulates seed development, germination, stem and root growth, cell division, and flowering time. It also enhances plant resilience to various environmental stresses, such as cold, drought, and heavy metal stress. Additionally, Gibberellic Acid has been observed to increase lipid peroxidation and cause fluctuations in the antioxidant defense system in rats. It is for research use only and not sold to patients.
  • Regulates plant development and growth
  • Promotes seed development and germination
  • Enhances stem and root growth
  • Supports cell division and flowering time
  • Improves plant response to environmental stresses like cold, drought, and heavy metals
  • Causes increased lipid peroxidation and affects antioxidant defense systems in rats
  • For research use only
Catalog No. 50-004-15216