Learn More
Alkali Scientific Gibberellic Acid A3 [GA3], 5 G
SDP
Supplier:  Alkali Scientific GIB002CP
Encompass_Preferred
Gibberellic Acid A3 (GA3) is a plant growth regulator in powder form, used for stimulating growth and promoting various biological activities in plants. It is widely used in laboratory applications to increase germination rates, promote stem elongation, and enhance fruit set in various plant species. As a bioactive compound, GA3 is often applied in controlled environments for research on plant physiology and growth regulation. It is an essential tool in agricultural research, plant breeding, and horticultural studies aimed at optimizing crop yield and improving plant growth under varying environmental conditions.
Catalog No. 50-006-60320