Learn More
Medchemexpress LLC Gibberellic acid (Synonyms: Gibberellin A3) | 77-06-5 | 346.37 | 5 G
SDP
Supplier:  Medchemexpress LLC HYN19645G
Encompass_Preferred
Gibberellic Acid, named after the fungus Gibberella fujikuroi, regulates processes in plant development and growth, including seed development and germination, stem and root growth, cell division, and flowering time. It also improves plant response to growth stress caused by various environmental stresses, such as cold stress, drought stress, and heavy metal stress. Additionally, it causes increased lipid peroxidation and fluctuations in the antioxidant defense system in rats.
  • Regulates plant development and growth
  • Supports seed development and germination
  • Enhances stem and root growth
  • Promotes cell division and flowering time
  • Improves plant response to environmental stresses like cold, drought, and heavy metals
Catalog No. 50-004-15071