Learn More
Medchemexpress LLC Giemsa stain | 51811-82-6 | 5 G
SDP
Supplier:  Medchemexpress LLC HYD09445G
Encompass_Preferred
Giemsa stain is a composite dye comprising methylene azure, methylene blue, eosin, and other components. It is utilized to stain chromatin, nuclear membranes, specific cytoplasmic components, and microorganisms.
  • Comprising methylene azure, methylene blue, eosin, and other components.
  • Utilized to stain chromatin.
  • Stains nuclear membranes.
  • Stains specific cytoplasmic components.
  • Stains microorganisms.
  • Intended for research in cytological and parasitological staining.
Catalog No. 50-004-10580