Learn More
Electron Microscopy Sciences Giemsa Stock Solution 250 ML
SDP
Supplier:  Electron Microscopy Sciences 2615303
Encompass_Preferred
Used in Differentiation of leucocytes, rickettsia, bacteria and inclusion bodies. Nuclei, additional leukocyte features are similar. References: Clark, G., Staining Procedures, Williams & Wilkins Co., Baltimore, 3rd Ed., p. 125, c. 1973
Catalog No. 50-316-98