Learn More
Polysciences, Inc. Gills modified EA, 1000 mL
SDP
Supplier:  Polysciences, Inc. 097831000
Encompass
Orange-6 and EA are the two cytoplasmic counterstain solutions that are used sequentially in the Papanicolaou staining method for clinical cytology. Gills modified OG-6 and EA are stable in solution and give predictable high quality staining results not previously possible with other formulations. Hazards: Flammable liquid , Irritant, Handling: Gloves & fume hood, Storage: Store at room temperature.
Catalog No. NC9002567