Learn More
Apexbio Technology LLC Gilteritinib 1254053-43-4 5mg
SDP
Supplier:  Apexbio Technology LLC A46915
Encompass_Preferred
Gilteritinib (CAS 1254053-43-4) also known as ASP2215 is a pyrazinecarboxamide derivative that functions as a selective inhibitor of FLT3 tyrosine kinase By targeting FLT3 gilteritinib disrupts aberrant signaling pathways associated with FLT3 mutations in hematologic malignancies while demonstrating minimal inhibition of c-Kit to reduce myelosuppressive effects observed with other FLT3 inhibitors Developed by Astellas Pharma gilteritinib is utilized in research focusing on FLT3-mutated acute myeloid leukemia and mechanisms of targeted kinase inhibition
Catalog No. 50-000-94078