Learn More
Medchemexpress LLC Gilteritinib-d8 | 2377109-74-3 | 98.0% | 560.76 | C29H36D8N8O3 | 1MG

Supplier: Medchemexpress LLC HY12432S11MG
Gilteritinib-d8 is the deuterium-labeled internal standard of gilteritinib (ASP2215), supplied as a solid analytical standard for research and quantitative mass spectrometry applications. It is intended for use in method development, ADME studies, and bioanalytical quantitation where an isotopically labeled reference is required.
- Deuterium-labeled internal standard for accurate mass spectrometry quantitation.
- High purity to minimize analytical interference.
- Molecular weight 560.76 and chemical formula C29H36D8N8O3 for unambiguous identification.
- Supplied as a solid, white to yellow, convenient for storage and handling.
- Recommended storage: powder -20°C for long-term; in solvent -80°C for extended stability.
- Suitable for ADME, pharmacokinetic, and method validation studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.