Learn More
Medchemexpress LLC HY-12432A 5mg Medchemexpress, Gilteritinib hemifumarate CAS:1254053-84-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12432A5MG
Encompass_Preferred
Medchemexpress, HY-12432A 5mg Gilteritinib hemifumarate CAS:1254053-84-3 Gilteritinib hemifumarate is a potent FLT3/AXL inhibitor with IC50 of 0.29 nM/0.73 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1043