Learn More
Selleck Chemical LLC Gilteritinib hemifumarate S4487-5mg
SDP
Supplier:  Selleck Chemical LLC S44875MG
Encompass_Preferred
Gilteritinib (ASP2215) hemifumarate is a potent and ATP-competitive FLT3/AXL inhibitor with IC50 of 0 29 nM/0 73 nM respectively
Catalog No. 50-311-0243