Learn More
GENOVIS INC GingisREX Lyophilized 5 x 5 ug, Lyophilized enzyme for arginine-specific protein digestion. Processes 5 x 0.1-1 mg protein.
Supplier:  GENOVIS INC B0GRX025
Encompass
Lyophilized enzyme for digestion C-terminally of arginine residues.GingisREX Lyophilized is available as a lyophilized powder in vials of 5 μg enzyme. Flexible format for method developmentPossible to combine enzymes for improved efficiencyAmenable to automated workflowsReady to use – just add water!
Catalog No. NC3762145